Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Tritylaniline 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-(Triphenylmethyl)aniline

A10550-14 A10550-14 A10550-06 A10550-06
AAA10550-06EA 45.99 USD InStock
CAAAA10550-14 CAAAA10550-06 AAA10550-06 AAA10550-14
4-Tritylaniline 97%
4-Tritylaniline


Formula: C₂₅H₂₁N
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00007898
CAS Number: 22948-06-7
EINECS: 245-346-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty