Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-3-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A10562-22 A10562-14 A10562-14 A10562-22
AAA10562-22EA 110.99 USD InStock
AAA10562-22 AAA10562-14 CAAAA10562-14 CAAAA10562-22
4-Hydroxy-3-nitrobenzoic acid 98%
4-Hydroxy-3-nitrobenzoic acid
Standard in chymotrypsin assays


Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00007119
CAS Number: 616-82-0
EINECS: 210-494-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty