Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromononane 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Nonyl bromide

A10577-14 A10577-14 A10577-36 A10577-22 A10577-36 A10577-22
AAA10577-22EA 124.99 USD InStock
AAA10577-22 AAA10577-14 AAA10577-36 CAAAA10577-22 CAAAA10577-14 CAAAA10577-36
1-Bromononane 99%
1-Bromononane


Formula: BrH₂C(CH₂)₇CH₃
MW: 207.15 g/mol
Boiling Pt: 200…201 °C
Melting Pt: –29 °C
Density: 1.086
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000278
CAS Number: 693-58-3
EINECS: 211-755-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty