Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isoniazid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Picolinic acid hydrazide 4-Pyridinecarboxylic acid hydrazide Isonicotinic acid hydrazide Pyridine-4-carboxylic acid hydrazide Isonicotinic hydrazide

A10583-18 A10583-0B A10583-36 A10583-36 A10583-22 A10583-22 A10583-18 A10583-0B
AAA10583-0BEA 200.99 USD InStock
AAA10583-0B CAAAAA10583-36 AAAA10583-22 CAAAA10583-18 AAAA10583-36 AAA10583-18 CAAAA10583-0B CAAAAA10583-22
Isoniazid 98+%
Isoniazid
Used for fluorescent HPLC detection of delta 4-3-ketosteroids


Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 169…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006426
CAS Number: 54-85-3
EINECS: 200-214-6
Merck Index: 13,05205
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty