Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Danger

A10585-0C A10585-36 A10585-0C A10585-36 A10585-0E A10585-0E
AAA10585-0CEA 314.99 USD InStock
CAAAA10585-0C CAAAA10585-36 CAAAA10585-0E AAA10585-0C AAA10585-36 AAA10585-0E
Nitrobenzene 99%
Nitrobenzene


Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 210…212 °C
Melting Pt: 5…6 °C
Density: 1.196
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007043
CAS Number: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: "6.1",II
Merck Index: 13,06621
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty