Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl bromoacetate 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bromoacetic methyl ester

A10605-18 A10605-0B A10605-0B A10605-18 A10605-30
AAA10605-30EA 124.99 USD InStock
AAA10605-30 CAAAA10605-0B AAA10605-18 AAA10605-0B CAAAA10605-18
Methyl bromoacetate 98+%
Methyl bromoacetate


Formula: BrCH₂COOCH₃
MW: 152.98 g/mol
Boiling Pt: 144…145 °C
Melting Pt: –50 °C
Density: 1.655
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000189
CAS Number: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty