Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3,5-diiodobenzoic acid tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

A10617-14 A10617-14 A10617-06 A10617-06
AAA10617-14EA 139.99 USD InStock
CAAAA10617-14 AAA10617-14 CAAAA10617-06 AAA10617-06
4-Amino-3,5-diiodobenzoic acid tech. 90%, Technical Grade
4-Amino-3,5-diiodobenzoic acid


Formula: C₇H₅I₂NO₂
MW: 388.93 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007684
CAS Number: 2122-61-4
EINECS: 218-331-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty