Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodine ≥99.9985% (metals basis), crystals, Puratronic®, resublimed

Supplier: Thermo Scientific Chemicals
Warning

010619-36 010619-88 010619-22 10619-14 010619-14
AAA10619-88EA 36 USD InStock
AAA10619-88 AAA10619-22 CAAA10619-14 AAA10619-36 AAA10619-14
Iodine ≥99.9985% (metals basis), crystals, Puratronic®, resublimed
Iodine


Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3 °C (1013 hPa)
Melting Pt: 183…186 °C (subl.)
Density: 4.930
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8,III
Merck Index: 13,05036
Order Now
Specification Test Results
Appearance (Color) Gray
Form Crystalline
Total Metal Impurities ≤0.0015% (15 ppm)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty