Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloropropanesulfonyl chloride ≥98%

Supplier: Thermo Scientific Chemicals
Danger

A10631-30 A10631-18 A10631-30 A10631-18 A10631-09 A10631-09
AAA10631-30EA 1284.99 USD InStock
AAA10631-30 CAAAA10631-30 AAA10631-09 AAA10631-18 CAAAA10631-09 CAAAA10631-18
3-Chloropropanesulfonyl chloride ≥98%
3-Chloropropanesulfonyl chloride


Formula: C₃H₆Cl₂O₂S
MW: 177.039 g/mol
Boiling Pt: 70 °C/0.5 mm
Density: 1.456
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007463
CAS Number: 1633-82-5
EINECS: 216-646-3
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty