Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Xylose 98+%

Supplier: Thermo Scientific Chemicals

A10643-22 A10643-0E A10643-0E A10643-22 A10643-36 A10643-36
AAA10643-0EEA 511.99 USD InStock
CAAAA10643-36 CAAAA10643-0E CAAAA10643-22 AAA10643-0E AAA10643-36 AAA10643-22
D-(+)-Xylose 98+%
D(+)-Xylose


Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 147…151 °C
Density: 1.525
Flash Pt: > 100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
EINECS: 200-400-7
Merck Index: 13,10142
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/28/2302128.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty