Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl (diethylphosphono)acetate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 2-(diethoxyphosphoryl)acetateDiethyl methoxycarbonylmethanephosphonate

A10644-36 A10644-22 A10644-36 A10644-14 A10644-14 A10644-22
AAA10644-22EA 287.99 USD InStock
CAAAA10644-36 CAAAA10644-14 AAA10644-22 CAAAA10644-22 AAA10644-36 AAA10644-14
Methyl (diethylphosphono)acetate 97%
Methyl (diethylphosphono)acetate


Formula: C₇H₁₅O₅P
MW: 210.17 g/mol
Boiling Pt: 126…128 °C/8 mm
Density: 1,145
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009081
CAS Number: 1067-74-9
EINECS: 213-938-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty