Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-L-leucine hydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-L-leucineBoc-Leu-OH

A10647-14 A10647-06 A10647-14 A10647-06
AAA10647-06EA 24.19 USD InStock
CAAAA10647-06 AAA10647-06 CAAAA10647-14 AAA10647-14
N-(tert-Butoxycarbonyl)-L-leucine hydrate 99%
N-(tert-Butoxycarbonyl)-L-leucine hydrate


Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol (anhy)
Melting Pt: 84…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066067
CAS Number: 13139-15-6
EINECS: 236-073-2
Order Now
Specification Test Results
Optical rotation -25° (c=2 in acetic acid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty