Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl decanoate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Capric acid ethyl ester Decanoic acid ethyl ester Decylic acid ethyl ester Ethyl caprate

A10657-0E A10657-22 A10657-0E A10657-22 A10657-36 A10657-36
AAA10657-22EA 32.69 USD InStock
CAAAA10657-36 AAA10657-22 CAAAA10657-0E AAA10657-0E CAAAA10657-22 AAA10657-36
Ethyl decanoate 99%
Ethyl decanoate


Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 250…252 °C
Melting Pt: –20 °C
Density: 0.862
Flash Pt: 102 °C(216 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty