Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(3-Aminopropyl)triethoxysilane 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-(Triethoxysilyl)propylamine Triethoxy(3-aminopropyl)silane Geniosil GF 93

A10668-22 A10668-36 A10668-22 A10668-36 A10668-0E A10668-0E
AAA10668-22EA 70.04 USD InStock
CAAAA10668-36 AAA10668-22 CAAAA10668-0E AAA10668-36 CAAAA10668-22 AAA10668-0E
(3-Aminopropyl)triethoxysilane 98%
(3-Aminopropyl)triethoxysilane


Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
Boiling Pt: 217 °C
Melting Pt: < –70 °C
Density: 0.948
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008207
CAS Number: 919-30-2
EINECS: 213-048-4
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty