Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus, red ≥99.999% (metals basis), lumps, Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

010670-18 010670-09 010670-03
AAA10670-09EA 173 USD InStock
AAA10670-09 AAA10670-18 AAA10670-03
Phosphorus, red ≥99.999% (metals basis), lumps, Puratronic®
Phosphorus


Formula: P
MW: 30.97 g/mol
Density: ∼2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133771
CAS Number: 7723-14-0
EINECS: 231-768-7
UN: 1338
ADR: 4.1,III
Order Now
Specification Test Results
Form Reddish-brown solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty