Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: o-Bromobenzonitrile

A10679-14 A10679-22 A10679-22 A10679-14 A10679-06 A10679-06
AAA10679-22EA 874.99 USD InStock
CAAAA10679-14 CAAAA10679-06 AAA10679-22 AAA10679-06 AAA10679-14 CAAAA10679-22
2-Bromobenzonitrile 99%
2-Bromobenzonitrile


Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 251…253 °C
Melting Pt: 53…56 °C
Density: 1.496
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001772
CAS Number: 2042-37-7
EINECS: 218-045-1
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty