Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Adamantanol 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Hydroxytricyclo[3.3.1.1(3,7)]decane 2-Tricyclo[3.3.1.1(3,7)]decanol Tricyclo[3.3.1.1(3,7)]decan-2-ol

A10697-14 A10697-06 A10697-06 A10697-14
AAA10697-06EA 49.81 USD InStock
AAA10697-06 AAA10697-14 CAAAA10697-06 CAAAA10697-14
2-Adamantanol 98%
2-Adamantanol


Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Melting Pt: 299…303 °C (sealed tube)
Density: 1.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074744
CAS Number: 700-57-2
EINECS: 211-846-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty