Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Thienylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Thiopheneacetic acidThiophene-3-acetic acid

A10700-22 A10700-22 A10700-14 A10700-06 A10700-14 A10700-06
AAA10700-22EA 676.99 USD InStock
AAA10700-22 AAA10700-14 CAAAA10700-06 CAAAA10700-14 AAA10700-06 CAAAA10700-22
3-Thienylacetic acid 98%
3-Thienylacetic acid


Formula: C₆H₆O₂S
MW: 142.18 g/mol
Boiling Pt: 129 °C/2.5 mm
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005473
CAS Number: 6964-21-2
EINECS: 230-166-1
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty