Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m-Anisic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Methoxybenzoic acid 3-Anisic acid

A10767-18 A10767-30 A10767-30 A10767-18
AAA10767-18EA 67.59 USD InStock
CAAAA10767-18 AAA10767-18 CAAAA10767-30 AAA10767-30
m-Anisic acid 99%
m-Anisic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 170…172 °C/10 mm
Melting Pt: 104…108 °C
Density: 1.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002499
CAS Number: 586-38-9
EINECS: 209-574-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty