Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Octylbenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A10777-03 A10777-06 A10777-06 A10777-03
AAA10777-06EA 281.99 USD InStock
CAAAA10777-06 CAAAA10777-03 AAA10777-06 AAA10777-03
4-Octylbenzoic acid 99%
4-Octylbenzoic acid


Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Boiling Pt: 362 °C (1013 hPa)
Melting Pt: 97…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00042649
CAS Number: 3575-31-3
EINECS: 222-692-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty