Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-L-tyrosine 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: BOC-L-TyrosineBoc-Tyr-OH

A10810-03 A10810-06 A10810-06 A10810-14 A10810-03 A10810-14
AAA10810-06EA 59.19 USD InStock
CAAAA10810-03 CAAAA10810-14 CAAAA10810-06 AAA10810-06 AAA10810-14 AAA10810-03
N-Boc-L-tyrosine 98+%
N-Boc-L-tyrosine


Formula: C₁₄H₁₉NO₅
MW: 281.3 g/mol
Melting Pt: 141 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00037179
CAS Number: 3978-80-1
EINECS: 223-613-7
Order Now
Specification Test Results
Optical rotation +37° (c=1 in dioxan)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty