Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Levulinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Oxopentanoic acid 4-Oxovaleric acid 4-Ketovaleric acid

A10813-30 A10813-0B A10813-30 A10813-0B
AAA10813-0BEA 129.99 USD InStock
AAA10813-0B AAA10813-30 CAAAA10813-30 CAAAA10813-0B
Levulinic acid 98%
Levulinic acid


Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 245…246 °C
Melting Pt: 26…31 °C
Density: 1.134
Flash Pt: 137 °C(278 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002796
CAS Number: 123-76-2
EINECS: 204-649-2
ADR: 8,III
Merck Index: 13,05492
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty