Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-4'-hydroxybiphenyl 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4'-Bromo-[1,1'-biphenyl]-4-ol

A10819-14 A10819-22 A10819-06 A10819-14 A10819-06 A10819-22
AAA10819-22EA 723.99 USD InStock
CAAAA10819-22 AAA10819-22 AAA10819-14 AAA10819-06 CAAAA10819-06 CAAAA10819-14
4-Bromo-4'-hydroxybiphenyl 98%
4-Bromo-4'-hydroxybiphenyl


Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Melting Pt: 163…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00059076
CAS Number: 29558-77-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty