Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1-Phenyl-1-propanol 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-α-Ethylbenzyl alcohol

A10830-14 A10830-22 A10830-36 A10830-22 A10830-14 A10830-36
AAA10830-22EA 66.09 USD InStock
AAA10830-22 CAAAA10830-14 AAA10830-36 AAA10830-14 CAAAA10830-36 CAAAA10830-22
(±)-1-Phenyl-1-propanol 98+%
(±)-1-Phenyl-1-propanol


Formula: C₆H₅CH(OH)CH₂CH₃
MW: 136.19 g/mol
Boiling Pt: 217…219 °C
Density: 0.994
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004564
CAS Number: 93-54-9
EINECS: 202-256-0
Merck Index: 13,03804
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/46/2299346.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty