Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Mannose 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

A10842-22 A10842-36 A10842-14 A10842-14 A10842-22 A10842-36
AAA10842-14EA 71.28 USD InStock
AAA10842-14 AAA10842-36 AAA10842-22 CAAAA10842-36 CAAAA10842-14 CAAAA10842-22
D-(+)-Mannose 99%
D(+)-Mannose


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 127…133 °C
Density: 1.54
Storage Temperature: Ambient
MDL Number: MFCD00064122
CAS Number: 3458-28-4
EINECS: 222-392-4
Merck Index: 13,05772
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty