Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Benzene sulphochloride

A10849-0I A10849-0B A10849-30 A10849-0I A10849-0B A10849-30
AAA10849-0BEA 95.39 USD InStock
AAA10849-0B CAAAA10849-0I CAAAA10849-30 AAA10849-0I CAAAA10849-0B AAA10849-30
Benzenesulfonyl chloride 98%
Benzenesulphonyl chloride


Formula: C₆H₅ClO₂S
MW: 176.62 g/mol
Boiling Pt: 252 °C
Melting Pt: 14…15 °C
Density: 1.377
Flash Pt: 132 °C(270 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: "8",III
Merck Index: 13,01072
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty