Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Phenylpropionyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrocinnamoyl chlorid 3-Phenylpropanoyl chloride 3-Phenylpropanonylchlorid

A10860-30 A10860-30 A10860-09 A10860-09
AAAA10860-30EA 759.99 USD InStock
AAAA10860-30 CAAAAA10860-30 CAAAA10860-09 AAA10860-09
3-Phenylpropionyl chloride 98%
3-Phenylpropionyl chloride


Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 225 °C
Melting Pt: –7 °C
Density: 1.135
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000748
CAS Number: 645-45-4
EINECS: 211-443-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty