Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isonicotinoyl chloride hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Pyridine-4-carbonyl Chloride Hydrochloride4-Pyridinoyl chloride hydrochloride

A10863-09 A10863-30 A10863-30 A10863-18 A10863-09 A10863-18
AAA10863-18EA 151.99 USD InStock
CAAAA10863-18 CAAAA10863-09 AAA10863-18 CAAAA10863-30 AAA10863-30 AAA10863-09
Isonicotinoyl chloride hydrochloride 98%
Isonicotinoyl chloride hydrochloride


Formula: C₆H₄ClNO
MW: 178.01 g/mol
Melting Pt: 154…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00012830
CAS Number: 39178-35-3
EINECS: 254-331-2
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty