Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Heptanesulfonic acid sodium salt, (max. 2% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 1-heptanesulfonate

A10917-22 A10917-22 A10917-14 A10917-06 A10917-14 A10917-06
AAA10917-22EA 695.32 USD InStock
AAA10917-22 CAAAA10917-06 CAA10917-22 CAA10917-14 AAA10917-06 AAA10917-14
1-Heptanesulfonic acid sodium salt, (max. 2% H₂O) 99% (dry weight)
1-Heptanesulphonic acid sodium salt
HPLC reagent used for ion-association in analyses of peptides and proteins


Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/91/2300791.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty