Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azomethine H sodium salt hydrate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate Azomethine-H monosodium salt hydrate

A10927-14 A10927-14 A10927-06 A10927-03 A10927-06 A10927-03
AAA10927-14EA 213.99 USD InStock
CAAAA10927-14 CAAAA10927-03 CAAAA10927-06 AAA10927-14 AAA10927-03 AAA10927-06
Azomethine H sodium salt hydrate
Azomethine H sodium salt hydrate


Formula: C₁₇H₁₂NNaO₈S₂·xH₂O
MW: 463.42 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149588
CAS Number: 206752-32-1
EINECS: 250-975-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty