Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-Hydroxyphenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Hydroxybenzyl cyanide

A10971-06 A10971-14 A10971-03 A10971-06 A10971-14 A10971-03
AAA10971-03EA 35.29 USD InStock
CAAAA10971-06 AAA10971-03 AAA10971-14 CAAAA10971-14 CAAAA10971-03 AAA10971-06
(4-Hydroxyphenyl)acetonitrile 97%
(4-Hydroxyphenyl)acetonitrile


Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 329…331 °C
Melting Pt: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002383
CAS Number: 14191-95-8
EINECS: 238-046-0
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty