Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrobromic acid 48% (w/w) in aqueous solution , 99,9999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrogen bromide

10991-AP 10991-AE 010991-AP 010991.AA 010991-AE 10991-AA
AA10991-APEA 1300 USD InStock
CAAA10991-AP AA10991-AP CAAA10991-AE AAA10991-AA AA10991-AE CAAAA10991-AA
Hydrobromic acid 48% (w/w) in aqueous solution , 99,9999% (metals basis)
Hydrogen bromide


Formula: HBr
MW: 80.91 g/mol
Density: 1.490
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II
Order Now
Specification Test Results
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty