Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Phenylbenzimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Phenyl-1H-benzimidazole

A10996-22 A10996-14 A10996-22 A10996-14 A10996-06 A10996-06
AAA10996-06EA 29.59 USD InStock
AAA10996-06 AAA10996-14 CAAAA10996-22 AAA10996-22 CAAAA10996-06 CAAAA10996-14
2-Phenylbenzimidazole 97%
2-Phenylbenzimidazole


Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
Boiling Pt: 422 °C (1013 hPa)
Melting Pt: 296…297 °C
Storage Temperature: Ambient
MDL Number: MFCD00005592
CAS Number: 716-79-0
EINECS: 211-939-2
Merck Index: 12,07428
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty