Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3'-Oxydipropionitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Bis(2-cyanoethyl) Ether

A11005-09 A11005-18 A11005-18 A11005-09 A11005-30 A11005-30
AAA11005-18EA 155.99 USD InStock
CAAAA11005-30 AAA11005-18 CAAAA11005-09 AAA11005-09 AAA11005-30 CAAAA11005-18
3,3'-Oxydipropionitrile 98%
3,3'-Oxydipropionitrile


Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 119…121 °C/1 mm
Density: 1.043
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001960
CAS Number: 1656-48-0
EINECS: 216-750-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty