Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α-Dimethoxytoluene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Benzaldehyde dimethyl acetal

A11009-22 A11009-22 A11009-36 A11009-36 A11009-0E A11009-0E
AAA11009-36EA 162.99 USD InStock
AAA11009-36 CAAAA11009-0E AAA11009-0E CAAAA11009-36 CAAAA11009-22 AAA11009-22
α,α-Dimethoxytoluene 99%
α,α-Dimethoxytoluene


Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 196 °C
Density: 1,014
Flash Pt: 69 °C(156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008491
CAS Number: 1125-88-8
EINECS: 214-413-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty