Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitro-L-tyrosine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-Tyr(3-NO2)-OH

A11018-14 A11018-06 A11018-22 A11018-06 A11018-22 A11018-14
AAA11018-22EA 589.99 USD InStock
AAA11018-22 CAAAA11018-14 CAAAA11018-06 CAAAA11018-22 AAA11018-06 AAA11018-14
3-Nitro-L-tyrosine 98%
3-Nitro-L-tyrosine


Formula: C₉H₁₀N₂O₅
MW: 226.3 g/mol
Melting Pt: ca 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007123
CAS Number: 621-44-3
EINECS: 210-688-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty