Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4-fluoroanisole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-4-fluoro-1-methoxybenzene

A11040-18 A11040-18 A11040-09 A11040-09
AAA11040-09EA 32.99 USD InStock
AAA11040-09 AAA11040-18 CAAAA11040-18 CAAAA11040-09
2-Bromo-4-fluoroanisole 98%
2-Bromo-4-fluoroanisole


Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 88…89 °C/1 mm
Melting Pt: –27 °C
Density: 1,581
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012014
CAS Number: 452-08-4
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty