Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-D-Lactose monohydrate

Supplier: Thermo Scientific Chemicals
Synonyms: α-Lactose monohydrate

A11074-0I A11074-0I A11074-30 A11074-0B A11074-0B A11074-30
AAA11074-0IEA 259.99 USD InStock
AAA11074-0I CAAAA11074-0I AAA11074-30 AAA11074-0B CAAAA11074-0B CAAAA11074-30
α-D-Lactose monohydrate
α-D-Lactose monohydrate


Formula: C₁₂H₂₄O₁₂
Melting Pt: 207…211 °C (dec.)
Density: 1,525
Storage Temperature: Ambient
MDL Number: MFCD00150747
CAS Number: 5989-81-1
EINECS: 200-559-2
Order Now
Specification Test Results
Optical rotation +52° (c=4.5 in water, 22h)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty