Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Vinylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A11107-14 A11107-14 A11107-06 A11107-06 A11107-03 A11107-03
AAA11107-06EA 387.99 USD InStock
AAA11107-06 AAA11107-14 AAA11107-03 CAAAA11107-14 CAAAA11107-03 CAAAA11107-06
4-Vinylbenzoic acid 98%
4-Vinylbenzoic acid


Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: ca 142 °C
Density: 1,30
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty