Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-fluorotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-4-fluoro-2-methylbenzene

A11112-22 A11112-14 A11112-14 A11112-22
AAA11112-22EA 141.99 USD InStock
CAAAA11112-22 AAA11112-22 AAA11112-14 CAAAA11112-14
2-Bromo-5-fluorotoluene 98+%
2-Bromo-5-fluorotoluene


Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 169…170 °C
Density: 1.495
Flash Pt: 70 °C(158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty