Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl formate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Formic acid ethyl ester

A11113-0F A11113-AP A11113-0F A11113-AE A11113-AE A11113-AP
AAA11113-APEA 70.53 USD InStock
AAA11113-AP CAAAA11113-AP AAA11113-AE AAA11113-0F CAAAA11113-0F CAAAA11113-AE
Ethyl formate 97%
Ethyl formate


Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 52…54 °C
Melting Pt: –81 °C
Density: 0.921
Flash Pt: -19 °C(-2 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: "3",II
Merck Index: 13,03841
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty