Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2,2-dimethoxypropane 98%

Supplier: Thermo Scientific Chemicals
Warning

A11139-09 A11139-18 A11139-30 A11139-09 A11139-18 A11139-30
AAA11139-18EA 185.99 USD InStock
AAA11139-18 AAA11139-09 CAAAA11139-30 CAAAA11139-09 AAA11139-30 CAAAA11139-18
1-Bromo-2,2-dimethoxypropane 98%
1-Bromo-2,2-dimethoxypropane


Formula: BrH₂CC(OCH₃)₂CH₃
MW: 183.05 g/mol
Boiling Pt: 83…87 °C/80 mm
Density: 1.354
Flash Pt: 40 °C(104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000208
CAS Number: 126-38-5
EINECS: 204-784-7
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty