Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-chlorobenzamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11153-06 A11153-06 A11153-03 A11153-03
AAA11153-03EA 50.29 USD InStock
AAA11153-03 AAA11153-06 CAAAA11153-03 CAAAA11153-06
2-Amino-5-chlorobenzamide 98+%
2-Amino-5-chlorobenzamide


Formula: C₇H₇ClN₂O
MW: 170.60 g/mol
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00017126
CAS Number: 5202-85-7
EINECS: 225-990-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty