Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Cyanobenzene Phenylcyanide

A11173-36 A11173-22 A11173-36 A11173-0E A11173-0E A11173-22
AAA11173-0EEA 154.99 USD InStock
CAAAA11173-36 CAAAAA11173-22 AAA11173-0E AAAA11173-22 AAA11173-36 CAAAA11173-0E
Benzonitrile 99%
Benzonitrile


Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 191…193 °C
Melting Pt: –13 °C
Density: 1.007
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001770
CAS Number: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: "6.1",II
Merck Index: 13,01098
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty