Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1,1'-Bi-2-naphthol 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,1'-Bi-2-naphthol2,2'-Dihydroxy-1,1'-binaphthyl1,1'-Binaphthyl-2,2'-Diol[1,1'-Binaphthalene]-2,2'-diol

A11182-14 A11182-14 A11182-06 A11182-22 A11182-06 A11182-22
AAA11182-06EA 46 USD InStock
CAAAA11182-22 AAA11182-06 CAAAA11182-14 AAA11182-14 CAAAA11182-06 AAA11182-22
(±)-1,1'-Bi-2-naphthol 99%
(±)-1,1'-Bi-2-naphthol


Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
Melting Pt: 215…218 °C
Density: 1.301
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 602-09-5
EINECS: 210-014-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty