Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Formylbenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Carboxybenzaldehyde Phthalaldehydic acid

A11201-18 A11201-0B A11201-30 A11201-30 A11201-18 A11201-0B
AAA11201-0BEA 787.99 USD InStock
CAAAA11201-18 AAA11201-0B AAA11201-30 CAAAA11201-0B AAA11201-18 CAAAA11201-30
2-Formylbenzoic acid 98+%
2-Formylbenzoic acid


Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 101 °C (0,4 mmHg)
Melting Pt: 97…101 °C
Density: 1,404
Flash Pt: 198 °C(388 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003336
CAS Number: 119-67-5
EINECS: 204-342-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty