Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Aminonicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Amino-3-pyridinecarboxylic acid

A11223-03 A11223-14 A11223-06 A11223-14 A11223-06 A11223-03
AAA11223-03EA 36.95 USD InStock
CAAAA11223-06 AAA11223-03 AAA11223-14 CAAAA11223-14 CAAAA11223-03 AAA11223-06
6-Aminonicotinic acid 98%
6-Aminonicotinic acid


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: ca 320 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006326
CAS Number: 3167-49-5
Merck Index: 13,00455
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty