Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Phenoxynicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Phenoxy-3-pyridinecarboxylic acid 2-Phenoxypyridine-3-carboxylic acid

A11247-03 A11247-06 A11247-06 A11247-03 A11247-14 A11247-14
AAA11247-06EA 71.79 USD InStock
CAAAA11247-06 AAA11247-06 AAA11247-14 AAAA11247-03 CAAAA11247-14 CAAAAA11247-03
2-Phenoxynicotinic acid 98%
2-Phenoxynicotinic acid


Formula: C₁₂H₉NO₃
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00014629
CAS Number: 35620-71-4
EINECS: 252-647-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty