Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Tartaric acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S,S)-(-)-Tartaric acid unnatural tartaric acid (S,S)-(-)-2,3-Dihydroxysuccinic acid

A11264-22 A11264-14 A11264-14 A11264-36 A11264-22 A11264-36
AAAA11264-36EA 412.99 USD InStock
AAAA11264-36 AAA11264-14 CAAAA11264-14 AAA11264-22 CAAAA11264-22 CAAAAA11264-36
D-(-)-Tartaric acid 99%
D(-)-Tartaric acid
Useful as a resolving agent


Formula: C₄H₆O₆
MW: 150.09 g/mol
Melting Pt: 166…170 °C
Flash Pt: 210 °C(410 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004238
CAS Number: 147-71-7
EINECS: 205-695-6
Merck Index: 12,09235
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty