Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Alanine ethyl ester hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: L-Ethyl alanate hydrochloride(S)-2-Aminopropionic Acid Ethyl Ester HydrochlorideEthyl (S)-2-Aminopropionate HydrochlorideH-Ala-OEt.HCl

A11271-06 A11271-14 A11271-14 A11271-06
AAA11271-14EA 107.99 USD InStock
CAAAA11271-06 AAA11271-14 AAA11271-06 CAAAA11271-14
L-Alanine ethyl ester hydrochloride ≥98%
L-Alanine ethyl ester hydrochloride


Formula: C₅H₁₁NO₂
MW: 153.61 g/mol
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00063662
CAS Number: 1115-59-9
EINECS: 214-225-9
Order Now
Specification Test Results
Optical rotation +2.9° (c=2.5 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty